Learn More
Cayman Chemical Meranzin, Purity: ≥98%, CAS: 23971-42-8, 10 mg
SDP
Catalog No. 502910257
Encompass_Preferred
A coumarin with diverse biological activities; cytotoxic to NSCLC-N6 and KB cancer cells (IC50s = 14.6 and 12.3 µg/ml, respectively); increases the latency to paw withdrawal in the hot plate test and reduces xylene-induced ear edema in mice at 10 mg/kg; decreases carrageenan-induced paw edema in rats at 10 mg/kg
Catalog No. 50-291-0257 Supplier Cayman Chemical Supplier No. 3474310 MG
A coumarin with diverse biological activities; cytotoxic to NSCLC-N6 and KB cancer cells (IC50s = 14.6 and 12.3 µg/ml, respectively); increases the latency to paw withdrawal in the hot plate test and reduces xylene-induced ear edema in mice at 10 mg/kg; decreases carrageenan-induced paw edema in rats at 10 mg/kg