Learn More
Medchemexpress LLC HY-10398 5mg Medchemexpress, CTS-1027 CAS:193022-04-7 Purity:>98%
SDP
Catalog No. 501968180
Encompass_Preferred
Medchemexpress, HY-10398 5mg CTS-1027 CAS:193022-04-7 CTS-1027 is a potent small molecule inhibitor of MMPs, with IC50s of 0.3 nM, 0.5 nM for MMP2, MMP13, respectively, and has > 1,000 fold selectivity over MMP1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8180 Supplier Medchemexpress LLC Supplier No. HY103985MG
Medchemexpress, HY-10398 5mg CTS-1027 CAS:193022-04-7 CTS-1027 is a potent small molecule inhibitor of MMPs, with IC50s of 0.3 nM, 0.5 nM for MMP2, MMP13, respectively, and has > 1,000 fold selectivity over MMP1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.