Learn More
Gibco™ CTS™ DPBS, calcium, magnesium, bag format
Description
Gibco™ Cell Therapy Systems (CTS) DPBS is the classical Dulbecco's phosphate buffered saline, which is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This version contains calcium and magnesium but no phenol red.
CTS quality
The Gibco™ CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485– and ISO 9001–certified. Gibco™ CTS products follow USP 'ancillary materials for cell, gene, and tissue-engineered products' within the responsibilities applicable to a supplier.
Gibco™ CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis, certificates of origin, and convenient access to our drug master file (DMF).
Closed systems compatible
This CTS DPBS (Cat. No. A4737901, 2 L) is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.
Specifications
Specifications
| Chemical Name or Material | DPBS |
| Concentration | 1X |
| For Use With (Application) | Cell and Gene Therapy Research, Development, and Manufacturing |
| Physical Form | Liquid |
| Quantity | 2 L |
| Solution Type | Dulbecco's Phosphate Buffered Saline |
| Packaging | BioProcess Container |
| Sterility | Sterile-filtered |
| Includes | Calcium and Magnesium |
| Manufacturing Quality | ISO13485, MDSAP, FDA-registered, 21 CFR820 |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.
Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.