Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ DPBS, without calcium chloride, without magnesium chloride

Catalog No. p-9277008
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1285602 2 L
A1285601 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1285602 Supplier Gibco™ Supplier No. A1285602
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.

Gibco Cell Therapy Systems (CTS) DPBS is the classic Dulbecco's phosphate-buffered saline. This particular formulation lacks calcium, magnesium, and phenol red.

CTS DPBS is manufactured in cGMP, ISO-certified facilities to help ensure the quality and consistency for reproducible results every day. Tests include osmolality, pH, stability, and the absence of bacterial/fungal contamination and endotoxin.

Gibco CTS products are high quality and supplied with harmonized documentation such as certificates of analysis, certificates of origin, and access to authorization letters for our Drug Master Files, as appropriate. The CTS product line enables you to reduce your burden when qualifying reagents during your transition from research to clinical applications.

Closed systems compatible

This CTS DPBS is provided in bioprocess container format with a chamber constructed from Aegis 5-14 film. The sterile, weldable DEPH-free PVC tubing with Luer lock and C-Flex™ tubing with MPC quick connector provide versatile connection options for closed system manufacturing processes.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DPBS
Concentration 1X
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Physical Form Liquid
Quantity 2 L
Solution Type Dulbecco's Phosphate Buffered Saline
Packaging Bag
Sterility Sterile-filtered
Includes No Calcium, No Magnesium
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21 CFR820
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.