Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ HBSS

Catalog No. A4000225403
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
2 L
Format:
Bag - BioProcess Container (BPC)
Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
A4000225403 2 L Bag - BioProcess Container (BPC)
A4000225401 1 L Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4000225403 Supplier Gibco™ Supplier No. A4000225403
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents.

Gibco Cell Therapy Systems (CTS) Hanks' Balanced Salt Solution (HBSS) is used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue samples, diluting cells for counting, and preparing reagents. This particular formulation lacks calcium, magnesium and phenol red.

Closed systems compatible
CTS HBSS is available in bottle and bioprocess container formats. The bioprocess container chamber is constructed from Aegis 5-14 film and includes sterile, weldable PVC/C-Flex hybrid tubing with a female Luer lock, providing versatile connection options for integrating into a closed system manufacturing process.

CTS quality
The Gibco CTS product line enables you to reduce your burden in qualifying reagents during your transition from research to clinical applications.

Products are manufactured at a site that uses methods and controls that conform to cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing sites are ISO 13485- and ISO 9001–certified. Gibco CTS products follow USP <1043> ancillary materials for cell, gene, and tissue-engineered products within the responsibilities applicable to a supplier.

Gibco CTS products are of high quality and are supplied with harmonized documentation such as certificates of analysis and certificates of origin.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15°C to 30°C
Culture Type Mammalian and Stem Cell Culture
Form Clear, colorless solution
Product Type Hanks' Balanced Salt Solution
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
Format Bag - BioProcess Container (BPC)
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO13485, MDSAP, FDA-registered, 21CFR820
Quantity 2 L
Shipping Condition Ambient
Show More Show Less

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.