Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ Opti-MEM™ I Medium

Catalog No. p-8750015
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4124801 100 mL
A4124802 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4124801 Supplier Gibco™ Supplier No. A4124801
Only null left
Add to Cart
Add to Cart

Gibco CTS Opti-MEM I Medium is an improved minimal essential medium (MEM) that is recommended for use with the CTS LV-MAX Transfection Kit as a complexation buffer.

Gibco CTS Opti-MEM I Medium is an improved minimal essential medium (MEM) that is recommended for use with the CTS LV-MAX Transfection Kit as a complexation buffer. This medium contains insulin, transferrin, hypoxanthine, thymidine, and trace elements. CTS Opti-MEM I uses a sodium bicarbonate buffer system (2.4 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH. The formulation of CTS Opti-MEM I is identical to research-grade Opti-MEM I Reduced Serum Medium.

CTS Opti-MEM I is xeno-free and contains human transferrin.

CTS Opti-MEM I is manufactured in conformity with cGMP for medical devices, 21 CFR, Part 820, and certified to the ISO 13485. Drug Master File and Regulatory Support File available upon request.

Custom configuration and packaging available upon request.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line Viral Production Cells
Cell Type HEK293
Classification Animal Origin-free
Form Liquid
Product Type Minimum Essential Medium (MEM)
Sterility Sterile-filtered
For Use With (Application) LV Production
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 100 mL
Shipping Condition Ambient
Format Bottle
Content And Storage 100 mL bottle. Store at 2°C to 8°C and protect from light.
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.