Learn More
Gibco™ CTS™ OpTmizer™ Pro SFM, bag format
Description
Gibco CTS OpTmizer Pro SFM, no phenol red, is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen specific) used in allogeneic cell therapies. This medium improves central memory phenotype and cell growth by shifting the cell's metabolism. The result is a larger population of central memory cells in a shorter period of time.
Benefits
- Maintains central memory T cells further into expansion and is well suited for longer workflows (>15 days)
- Offers >90% (average) T cell fold expansion (from healthy donors) compared to CTS OpTmizer medium
- Modest increases in central memory cells at early stage in expansion protocol result in larger cell yields at harvest
- Helps achieve the same number of cells in a shorter duration
- Ideally suited to the production of allogeneic therapies
Features
- Serum-free and xeno-free
- Supports high density cultures in static cultures
- New film (Aegis) and bag design allow compatibility with closed systems
- Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols
Increased efficiency and consistence performance
CTS OpTmizer Pro SFM, no phenol red, is serum-free and xeno-free and supports high density cultures in static cultures. It displays consistent performance in the support of T-cell expansion with better central memory phenotype in a shorter time.
Enhanced expansion
CTS OpTmizer Pro SFM, no phenol red, increases the number of central memory cells at an early stage in the expansion process and maintains T-cell functionality.
Enhanced design and compatibility
Use of Thermo Scientific Aegis5-14 Film allows compatibility with closed systems. This film is a five-layer cast film produced in a cGMP facility. The outer layer is a polyester elastomer coextruded with an ethyl vinyl alcohol (EVOH) barrier layer and an ultra-low density polyethylene product contact layer. Aegis5-14 Film is manufactured using no animal derived components.
Simplify your transition into clinical research
The Gibco Cell Therapy Systems (CTS) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:
- Reduced burden in qualifying reagents
- Appropriate certificates of analysis and origin available
- Convenient access to our Drug Master File (DMF)
Specifications
Specifications
| Cell Type | T Cells |
| Classification | Xeno-free |
| Product Type | Serum Free Medium (SFM) |
| Serum Level | Serum-free |
| Sterility | Sterile |
| Without Additives | No Glutamine, No Phenol Red |
| Quantity | 1 L |
| Culture Type | Static Culture |
Frequently Asked Questions (FAQs)
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.