Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ OpTmizer™ Pro SFM, no phenol red

Catalog No. A4966105
Change view
Click to view available options
Quantity:
5 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4966105 5 L
A4966107 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A4966105 Supplier Gibco™ Supplier No. A4966105
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies.

Gibco™ CTS OpTmizer Pro SFM is a novel serum-free medium (SFM) developed for the growth and expansion of human T lymphocytes (e.g., CD4+, CD8+, polyclonal, or antigen-specific) used in allogeneic cell therapies. This medium improves central memory phenotype and cell growth by shifting the cell's metabolism. The result is a larger population of central memory cells in a shorter period of time.

CTS OpTmizer Pro SFM consists of CTS OpTmizer Pro Basal Medium and CTS OpTmizer Supplement, which are mixed together prior to use.

CTS OpTmizer Pro SFM in 5 L and 10 L bag format is compatible with closed-system processing for media preparation and integration to standard cell therapy equipment, including CTS devices (Xenon, Rotea, and DynaCellect).

Benefits of CTS OpTmizer Pro SFM include:

  • Supplement component now in bag format to allow for closed mixing with basal medium
  • Reduces the number of open steps in your process, thus reducing contamination risks
  • Easy integration with cell therapy equipment, including CTS Xenon, Rotea, and DynaCellect
  • Maintains central memory T cells further into expansion and is well suited for longer workflows (>15 days)
  • Offers >90% (average) T-cell fold-expansion (from healthy donors) compared to CTS OpTmizer medium
  • Modest increases in central memory cells at early stage in expansion protocol result in larger cell yields at harvest
  • Helps achieve the same number of cells in a shorter duration
  • Ideally suited to the production of allogeneic therapies

Features include:

  • Flexible pack sizes in Aegis5-14 film: basal medium in 5 L and 10 L bags
  • Weldable C-Flex and PVC lines
  • Supports allogeneic T-cell processes through all stages of development and manufacturing
  • Supports T-cell activation using Dynabeads magnetic beads, soluble antibodies, and stimulatory antibody-presenting cell protocols

Increased efficiency and consistence performance
CTS OpTmizer Pro SFM, no phenol red, is serum-free and xeno-free and supports high density cultures in static cultures. It displays consistent performance in the support of T-cell expansion with better central memory phenotype in a shorter time.

Enhanced expansion
CTS OpTmizer Pro SFM, no phenol red, increases the number of central memory cells at an early stage in the expansion process and maintains T-cell functionality.

Enhanced design and compatibility
The supplement component is now in a bag with a weldable PVC line, allowing for closed-system mixing with the CTS OpTmizer Pro basal medium bag. This feature, along with larger 5 L and 10 L basal medium sizes, addresses manufacturers' need for scale, efficiency, and contamination risk mitigation.

Use of Thermo Scientific™ Aegis5-14 Film allows compatibility with closed systems. This film is a five-layer cast film produced in a cGMP facility. The outer layer is a polyester elastomer coextruded with an ethyl vinyl alcohol (EVOH) barrier layer and an ultra-low density polyethylene product contact layer. Aegis5-14 Film is manufactured using no animal derived components.

Simplify your transition into clinical research
The Gibco™ Cell Therapy Systems (CTS) brand offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices.

CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)

Specifications

Cell Type T Cells
Classification Xeno-free
Form Liquid
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method 0.2 μm Sterile-filtered
With Additives None
Without Additives No Phenol Red
Product Line OpTmizer™ Pro
Quantity 5 L
Shipping Condition Room Temperature
Content And Storage • 1 x 5000 mL bag CTS OpTmizer Pro Basal Medium, store at 2°C to 8°C, protect from light
• 1 x 130 mL CTS OpTmizer Expansion Supplement in 250 mL bag, store at 2°C to 8°C, protect from light
Culture Environment Bags, Bioreactors
Culture Type Static Culture
Show More Show Less
Are the 5L and 10L sizes/configurations of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) compatible with CTS devices?

Yes. The 5L and 10L BPCs (bioprocess containers) are weldable to Xenon, Rotea, and DynaCellect consumables.

With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how much supplement should be added to the basal medium?

The 5L and 10L kits are designed so that the entire volume of the supplement BPC (bioprocess container) is added to the basal medium BPC.

With the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107), how is the supplement added to the basal medium?

The supplement BPC (bioprocess container) has 24 in. of sterile weldable 3/32 in. ID, 5/32 in. OD DHEP-free PVC tubing. With fill volumes of 130 mL and 260 mL for the 5L and 10L kits respectively, the supplement BPC can be directly welded to the basal medium BPC.

What are the differences between the BPCs (bioprocess containers) in the 1L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM vs. the 5L and 10L sizes?

In the 5L and 10L sizes of CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM, both the basal medium and supplement components are in BPCs (bioprocess containers) composed of Aegis 5-14 film. The basal medium BPC has PVC and C-Flex lines whereas the supplement BPC has a PVC line.

What is the stability of the CTS OpTmizer SFM, no phenol red and CTS OpTmizer Pro SFM in the new 5L and 10L sizes?

The 5L and 10L sizes of both CTS OpTmizer SFM, no phenol red (Cat. No. A3705005, A3705007) and CTS OpTmizer Pro SFM (Cat. No. A4966105, A4966107) have 12-month expiry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.