Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ CTS™ PSC Cryomedium

Catalog No. A4238801
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4238801 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4238801 Supplier Gibco™ Supplier No. A4238801
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CTS PSC Cryomedium is a cGMP-manufactured, ready-to-use, xeno-free solution for the cryopreservation of early passage pluripotent stem cells (PSCs).

CTS PSC Cryomedium is a cGMP-manufactured, ready-to-use, xeno-free solution for the cryopreservation of early passage pluripotent stem cells (PSCs). It helps minimize loss of cell viability and maximize post-thaw recovery. For the cryopreservation and recovery of PSCs, use the CTS PSC Cryopreservation Kit, which includes CTS PSC Cryomedium and CTS RevitaCell Supplement.

CTS PSC Cryomedium:

  • Provides efficient recovery and higher viability than traditional methods of PSC cryopreservation
  • Facilitates regulatory filings through cGMP manufacturing and the availability of documentation
  • Simplifies the transition from bench to clinic through the direct replacement of RUO PSC Cryopreservation Medium

Efficient recovery

CTS PSC Cryomedium routinely achieves higher viability than traditional methods of cryopreservation and is compatible with clump and single cell passaged PSCs cultured in a variety of media systems. It is provided as a ready-to-use, complete solution containing 10% DMSO for efficient cell addition. Separate additions of enhancing reagents are not required, helping save valuable preparation time.

Facilitates regulatory filings

CTS PSC Cryomedium is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability and convenient access to our Drug Master File (DMF) are available for CTS PSC Cryomedium.

Simplifies transition from bench to clinic

With its extensive safety testing, including adventitious agents, CTS PSC Cryomedium enables a smooth transition from RUO PSC Cryopreservation Medium.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs)
Classification Xeno-free
Form Liquid
Product Type Cryopreservation Medium
Sterility Sterile-filtered
With Additives DMSO (10%)
For Use With (Application) Cell and Gene Therapy Research, Development, and Manufacturing
Manufacturing Quality ISO 13485, MDSAP, FDA-registered, 21 CFR 820
Quantity 50 mL
Shipping Condition Dry Ice
Format Bottle
Content And Storage Store at –5 to –20°C and protect from light.
Show More Show Less
What is CTS?

The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems.

What is the difference between the PSC Cryopreservation Kit and the CTS PSC Cyropreservation Kit?

The CTS PSC Cryopreservation Kit (Cat. No. A4239301) provides the same efficient recovery, consistency, and reduced variability as the PSC Cryopreservation Kit (Cat. No. A2644601). However, these two kits have different intended use statements, i.e., “For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals” and “For Research Use Only”, respectively. Furthermore, the CTS PSC Cryopreservation Kit has full USP sterility testing info and a Drug Master file (DMF), whereas the sterility testing of the PSC Cryopreservation is via a modified USP method, and it does not have a DMF.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.