Learn More
Cole-Parmer Oakton Replacement Sensor for CTSTestr 5
Supplier:  Cole-Parmer 3563444
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample. For your convenience, cables, power adapters, and other accessories are available for your Oakton meters and pocket testers.
Catalog No. 50-196-1768