Learn More
Aviva Systems Biology Corp CTSC Antibody - middle region (ARP60116_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60116_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CTSC. Alternate names: CPPI, DPP1, DPPI, HMS, JP, JPD, PALS, PLS, DPP-IApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse
Catalog No. 50-286-945