Learn More
Aviva Systems Biology Corp Ctsd Antibody - C-terminal region (ARP41481_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41481_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ctsd. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: CD, CatDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-284-936