Learn More
Aviva Systems Biology Corp CTSE Antibody - N-terminal region (ARP60124_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60124_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSE. It was validated on Western Blot. . Alternate names: CATEApplicationWB. Species reactivity: Horse, Human
Catalog No. 50-286-950