Learn More
Aviva Systems Biology Corp CTSG Antibody - C-terminal region (ARP61136_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61136_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSG. It was validated on Western Blot. . Alternate names: CG, MGC23078, CATGApplicationWB. Species reactivity: Guinea Pig, Human, Rat
Catalog No. 50-287-265