Learn More
Aviva Systems Biology Corp CTSH Antibody - C-terminal region (ARP63370_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63370_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSH. It was validated on Western Blot. . Alternate names: ACC-4, ACC-5, CPSB, DKFZp686B24257, MGC1519, minichainApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-950