Learn More
Aviva Systems Biology Corp CTSK Antibody - middle region (ARP60949_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60949_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSK. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CTSK. Alternate names: CTS02, CTSO, CTSO1, CTSO2, MGC23107, PKND, PYCDApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-209