Learn More
Aviva Systems Biology Corp CTSL2 Antibody - N-terminal region (ARP61285_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61285_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSL2. It was validated on Western Blot. . Alternate names: CATL2, CTSU, CTSV, MGC125957, CTSL2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-327