Learn More
Aviva Systems Biology Corp CTSL2 Antibody - C-terminal region (ARP61284_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61284_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CTSL2. It was validated on Western Blot. . Alternate names: CATL2, CTSU, CTSV, MGC125957, CTSL2ApplicationWB. Species reactivity: Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-326