Learn More
Enzo Life Sciences CTT (1mg)
SDP
Supplier:  Enzo Life Sciences BMLPI1360001
Encompass_Preferred
CTTHWGFTLC is a specific inhibitor of the gelatinases, MMP-2 and MMP-9, with an IC50 of 10-50 µM. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo. Also available, negative control peptide STTHWGFTLS. Alternative name: Gelatinase inhibitor. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water at 1mg/ml. Long Term Storage: -20°C.
Catalog No. 50-201-1473