Learn More
SKYLINE CENTER Item #W82 Bag 7/8” Round - 500ct.
Catalog No. NC0829132
Encompass
7/8” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - .17 - .22 Caliber, .22 - .25 Caliber
Catalog No. NC0829132 Supplier SKYLINE CENTER Supplier No. 82/RC
7/8” ROUND 100% COTTON PATCHES Uses: Cleaning - guns, firearms, melt flow index, melt plastometers, capillary rheometers, optics, laboratory equipment. Gun Cleaning - .17 - .22 Caliber, .22 - .25 Caliber