Learn More
Cayman Chemical CTX1211trIfluoroACTAte sa 10mg
SDP
Catalog No. 502943123
Encompass_Preferred
A peptide agonist of melanocortin receptors; increases cAMP levels in cells expressing MC1R, MC3R, or MC4R (EC50s = 6.2, 3.9, and 2.7, nM, respectively); sensitizes male and female mice to liraglutide-induced reductions in food intake and body weight at 1 mg/kg
Catalog No. 50-294-3123 Supplier Cayman Chemical Supplier No. 4208110 MG
A peptide agonist of melanocortin receptors; increases cAMP levels in cells expressing MC1R, MC3R, or MC4R (EC50s = 6.2, 3.9, and 2.7, nM, respectively); sensitizes male and female mice to liraglutide-induced reductions in food intake and body weight at 1 mg/kg