Learn More
TARGETMOL CHEMICALS INC CU-115 5MG
SDP
Catalog No. 502578987
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CU-115 is a selective and potent TLR8 antagonist with IC50 of 1.04 µM and =>50 µM for TLR8 and TLR7 respectively. CU-115 decreases production of TNF-(alpha) and IL-1(beta) activated by R-848 in THP-1 cells. purity: 100%
Catalog No. 50-257-8987 Supplier TARGETMOL CHEMICALS INC Supplier No. T96455MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CU-115 is a selective and potent TLR8 antagonist with IC50 of 1.04 µM and =>50 µM for TLR8 and TLR7 respectively. CU-115 decreases production of TNF-(alpha) and IL-1(beta) activated by R-848 in THP-1 cells. purity: 100%