Learn More
Bioworld CU-CHELATED SEPAROPOREAGA
Catalog No. 50197787
Encompass
CU-CHELATED SEPAROPORE (AGAROSE): Cu-Chelated Separopore is an imminodiacetate (IDA) immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity to Cumetal ions. Properties: Affinity...
Catalog No. 50-197-787 Supplier Bioworld Supplier No. 201810481
CU-CHELATED SEPAROPORE (AGAROSE): Cu-Chelated Separopore is an imminodiacetate (IDA) immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity to Cumetal ions. Properties: Affinity...