Learn More
Bioworld CU-CHELATED SEPAROPOREAGA
Supplier:  Bioworld 201810482
Encompass
CU-CHELATED SEPAROPORE (AGAROSE): Cu-Chelated Separopore is an imminodiacetate (IDA) immobilized metal affinity chromatography (IMAC) matrix that seperates proteins via their differences in affinity to Cumetal ions. Properties: Affinity...
Catalog No. 50-197-788