Learn More
Strem, An Ascensus Company CAS# 149-11-1. 100g. Copper(II) 2-ethylhexanoate (solvent free - 16-19% Cu). MFCD00015695
SDP
Supplier:  Strem, An Ascensus Company 292941100G
Encompass
CAS# 149-11-1. 100g. Copper(II) 2-ethylhexanoate (solvent free - 16-19% Cu). MFCD00015695. Molecular Weight: 349.96. Molecular Formula: Cu[OOCCH(C2H5)C4H9]2. Color/form: blue pwdr. Strem# 29-2941. http://www.strem.com/catalog/v/29-2941/
Catalog No. 50-201-6806