Learn More
Echelon Biosciences Research Labs CU(II) PROTOPORPHYRIN IX 50 mg
SDP
Supplier:  Echelon Biosciences Research Labs FH07050MG
Encompass
Cu (II) Protoporphyrin IX does not inhibit heme oxygenase (the enzyme that catalyzes the conversion of heme to biliverdin in the heme degradation pathway) and is used as a negative control for Zn (II) Protoporphyrin (an inihibitor of heme oxygenase). Heme oxygenase has been implicated in tumor cell resistance to chemotherapy, reduction of free radical formation and inflammation, and associated with vascular repair. This item is non returnable
Catalog No. 50-720-334