Learn More
Bioworld Cu-IMAC 5 x 1mL Columns, 5 Column
Supplier:  Bioworld 201700081
Encompass_Preferred
Cu-immobilized metal affinity chromatography (IMAC) columns consist of Cu-Chelated Separoporematrix conveniently prepacked spin columns for gravity flow chromatography.Cu-IMAC Columns separate proteins via their differences in affinity toCu2+ metal ions. Ideal for purification of recombinant proteins containing (His)6 fusion tag.Technical SpecificationsImminodiacetate immobilized matrix charged withCu metal ions.Chelating group coupling method: epoxy activationMatrix: cross-linked Separopore 4BParticle size (range): 50-170µm Metal ion capacity: > 20µmol Cu2+/ml gelSpacer arm: 14 atompH stability: 3-12Binding Capacity: Approx. 40 mg His-tagged protein/ml mediumSupplied as suspension in 20% ethanol.
Catalog No. 50-197-628