Learn More
AccuStandard, Inc CU STD IN HNO3, 100ML
SDP
Catalog No. NC0955430
Encompass
Copper ICP Standard. 5 x 1 mL. 2.5 mg/mL in MeOH.
Catalog No. NC0955430 Supplier AccuStandard, Inc Supplier No. ICP15N1
Copper ICP Standard. 5 x 1 mL. 2.5 mg/mL in MeOH.