Learn More
Medchemexpress LLC HY-110353 5mg Medchemexpress, CU-T12-9 CAS:1821387-73-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1103535MG
Encompass_Preferred
Medchemexpress, HY-110353 5mg CU-T12-9 CAS:1821387-73-8 CU-T12-9 is a specific TLR1/2 agonist with EC50 of 52.9 nM in HEK-Blue hTLR2 SEAP assay. CU-T12-9 activates both the innate and the adaptive immune systems. CU-T12-9 selectively activates the TLR1/2 heterodimer, not TLR2/6. CU-T12-9 signals through NF-κB and invokes an elevation of the downstream effectors TNF-α, IL-10, and iNOS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8794