Learn More
Enzo Life Sciences Cu/Zn SOD polyclonal antibody (100 µg)
SDP
Supplier:  Enzo Life Sciences ADISOD101E
Encompass_Preferred
Host: Rabbit. Immunogen: Native rat Cu/Zn SOD. UniProt: P07632. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat. Applications: WB. Recommended dilutions: Western Blot (0.5µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-407