Learn More
Enzo Life Sciences Cu/Zn SOD polyclonal antibody (1mg)
SDP
Supplier:  Enzo Life Sciences ADISOD100J
Encompass_Preferred
Host: Rabbit. Immunogen: Native human Cu/Zn SOD. UniProt ID: P00441. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Clam. Applications: IF, IHC (PS), IP, WB. Recommended Dilutions: Immunofluorescence (1:50-1:200) Immunohistochemistry (paraffin sections, 1:50) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7406