Learn More
Enzo Life Sciences Cu/Zn SOD polyclonal antibody (biotin conjugate) (200µg)
SDP
Supplier:  Enzo Life Sciences ADISOD100B0200
Encompass_Preferred
Host: Rabbit. Immunogen: Native human Cu/Zn SOD. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat Clam. Applications: IF, IHC (PS), IP, WB. Purity: Protein A-affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7405