Learn More
Enzo Life Sciences Cu/Zn SOD polyclonal antibody (50 µg)
SDP
Supplier:  Enzo Life Sciences ADISOD100D
Encompass_Preferred
Host: Rabbit. Immunogen: Native human Cu/Zn SOD. UniProt: P00441. Source: Purified from rabbit serum. Species Reactivity: Human, Mouse, Rat, Clam. Applications: IF, IHC (PS), IP, WB. Recommended dilutions: Immunofluorescence (1:50-1:200). Immunohistochemistry (paraffin sections, 1:50). Western Blot (1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-667-404