This product consists of pre-assembled nanodiscs incorporating MSP1D1 dH5 membrane scaffold proteins and POPC phospholipids. Designed for the stabilization of cell-free expressed membrane proteins, these nanodiscs provide a native-like lipid environment by removing proteins from the cell membrane. The construct features an N-terminal His-tag to facilitate purification and immobilization.
- Pre-assembled format for immediate use in membrane protein studies
- Constructed with MSP1D1 dH5 scaffold protein for approximately 7-8 nm diameter particles
- Includes N-terminal His-tag for affinity-based applications
- Supplied in 20 mM TRIS pH 7.4, 100 mM NaCl, and 0.5 mM EDTA buffer
- Purity level greater than 90%
- Molecular mass of 21,468 Da
- Recommended storage at -80°C to avoid repeated freeze-thaw cycles