Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genomadix™ Cube CYP2C19 External Control Cartridges

Catalog No. 22040048
Change view
Click to view available options
Clia Complexity:
High
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-040-048 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-040-048 Supplier Genomadix™ Supplier No. ASM00099
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cube CYP2C19 External Control Cartridges are cartridges that contain required material to run an external control test on the Genomadix Cube system. Quantity: 10 Control Tests.

  • Cartridges are run to determine if the Genomadix Cube is performing correctly.
  • Cartridges contain synthetic DNA template and the necessary reagents for DNA extraction, PCR amplification and detection of CYP2C19 alleles.
  • Cartridges do not require any additional contents to be added.
  • Compatible with the FDA 510(k)-cleared Genomadix Cube CYP2C19 System.

Specifications

Clia Complexity High
Type External Control Cartridge
For Use With (Application) Testing the performance of the Genomadix Cube
Form Frozen

FDA 510(K)-cleared

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.