Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genomadix™ Cube CYP2C19 Swab Kit

Catalog No. 22040049
Change view
Click to view available options
Clia Complexity:
High
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-040-049 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-040-049 Supplier Genomadix™ Supplier No. ASM00103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cube CYP2C19 Swab Kit is a pouch containing 3 swabs, used to collect buccal samples from the inside of a patient’s cheek. Quantity: 20 pouches (3 swabs/pouch)

  • Proprietary swabs used for collection of buccal samples from the inside of a patient's cheek and transferred to the cartridge tubes.
  • Swab pouches contain 3 swabs.
  • Label on the pouch contains fields to track information including patient ID, swab collector, date, and time.
  • Compatible with the FDA 510(k)-cleared Genomadix Cube CYP2C19 System.

Specifications

Clia Complexity High
Type Swab Kit
For Use With (Application) Collecting a buccal sample for the Genomadix CYP2C19 System
Form Swab

FDA 510(K)-cleared

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.