Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genomadix™ Cube CYP2C19 Test Kit

Catalog No. 22040047
Change view
Click to view available options
Clia Complexity:
High
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-040-047 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-040-047 Supplier Genomadix™ Supplier No. ASM00095
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cube CYP2C19 Test Kits contain test cartridges and swab kits, used to detect a patient's CYP2C19 2, 3, and 17 alleles. Quantity: 20 tests.

  • Test Cartridges contain the reagents needed for DNA extraction, PCR amplification and detection of CYP2C19 alleles.
  • The CYP2C19 2, 3, and 17 alleles are detected in three separate tubes specific for each allele.
  • The presence or absence of genotypes are determined using Polymerase Chain Reaction (PCR) and fluorescent probe detection.
  • Swab kits are used to collect buccal material from a patient for analysis in the test cartridges
  • Compatible with the FDA 510(k)-cleared Genomadix Cube CYP2C19 System.

Specifications

Clia Complexity High
Type Test Kit
For Use With (Application) Genotype Testing
Form Frozen

FDA 510(K)-cleared

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.