Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genomadix™ Cube CYP2C19 Training Kit

Catalog No. 22040050
Change view
Click to view available options
Clia Complexity:
High
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Clia Complexity
22-040-050 High
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-040-050 Supplier Genomadix™ Supplier No. ASM00116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Cube CYP2C19 Training Kit contains cartridges filled with water and swab pouches to be used for training purposes.

  • The training kit contains a combination of swab pouches and cartridges containing water for training purposes.
  • Used in conjunction with the Training Module available within the Genomadix Cube CYP2C19 system.
  • Compatible with the FDA 510(k)-cleared Genomadix Cube CYP2C19 System.

Specifications

Clia Complexity High
Type Training Kit
For Use With (Application) Training users how to operate the Genomadix CYP2C19 System
Form Liquid Ready-to-Use

FDA 510(K)-cleared

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.