Learn More
Andwin Scientific CUBE RACKS/82010-746 5/PK
SDP
Catalog No. NC9411310
Encompass_Preferred
The cube racks provide support for four 50 mL tubes, ten 15 mL conical tubes, twelve 12x75mm or 12x100mm tubes, or sixteen 1.52.0 mL microcentrifuge tubes. Each cube rack has four sides. This product is manufactured from polypropylene. It is sold in a pack of 5 cube racks in assorted colors of blue, green, pink, yellow, and orange.
Catalog No. NC9411310 Supplier Andwin Scientific Supplier No. 82010746
The cube racks provide support for four 50 mL tubes, ten 15 mL conical tubes, twelve 12x75mm or 12x100mm tubes, or sixteen 1.52.0 mL microcentrifuge tubes. Each cube rack has four sides. This product is manufactured from polypropylene. It is sold in a pack of 5 cube racks in assorted colors of blue, green, pink, yellow, and orange.