Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCI America™ CUBIC-HV™ 3D Nuclear Staining Kit
SDP

Catalog No. C37091KIT Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C37091KIT 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C37091KIT Supplier TCI America™ Supplier No. C37091KIT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This kit contains buffers and anti-NeuN antibody (positive control) for 3D nuclear staining of CUBIC-cleared tissues. It is suitable for the use on the brain of an adult mouse. Please note that CUBIC-L (T3740) and CUBIC-R+(M) (T3741) are required to clear tissue first.

This kit contains the following:

  • CUBIC-HV™1 3D nuclear staining buffer (1x) (for 10 tests)
  • CUBIC-HV™1 3D nuclear staining washing buffer (100x) (for 10 tests)
  • DAPI·2HCl (200x) (for 10 tests)
  • Propidium Iodide (100x) (for 10 tests)
  • 10 x 5 mL tube

Specifications

For Use With (Equipment) Light-sheet Fluorescent Microscopy (LSFM) or Confocal Laser-scanning Microscopy (CLSM) Device
Product Line CUBIC
Product Type Special Stain
Quantity 1 Kit
For Use With (Application) Neuroscience, 3D pathology, oncology, histology, immunology, and other research applications
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.