Learn More
Medchemexpress LLC Cucurbit[7]uril | 259886-50-5 | MFCD03456500 | 98.0% | 1,162.96 g·mol⁻1 | C42H42N28O14 | 250 MG
SDP
Catalog No. 5000243540
Encompass_Preferred
Cucurbit[7]uril is a cyclic macrocycle made of seven glycoluril units linked by methylene bridges, forming a rigid, barrel-shaped cavity that selectively encapsulates appropriately sized guest molecules. It is widely used in supramolecular chemistry and biochemical research for host-guest investigations, sensing, catalysis, and studies of molecular recognition and delivery.
  • Macrocyclic host with a rigid barrel-shaped cavity.
  • Selective encapsulation of guest molecules based on size and polarity.
  • High binding affinity for a range of organic and inorganic guests.
  • Common applications include host-guest studies, sensing, and catalysis.
  • Useful for drug delivery research and formulation studies.
Catalog No. 50-002-43540 Supplier Medchemexpress LLC Supplier No. HY104086250MG
Cucurbit[7]uril is a cyclic macrocycle made of seven glycoluril units linked by methylene bridges, forming a rigid, barrel-shaped cavity that selectively encapsulates appropriately sized guest molecules. It is widely used in supramolecular chemistry and biochemical research for host-guest investigations, sensing, catalysis, and studies of molecular recognition and delivery.
  • Macrocyclic host with a rigid barrel-shaped cavity.
  • Selective encapsulation of guest molecules based on size and polarity.
  • High binding affinity for a range of organic and inorganic guests.
  • Common applications include host-guest studies, sensing, and catalysis.
  • Useful for drug delivery research and formulation studies.