Learn More
Cayman Chemical CucurbItacIn B 10mg
SDP
Catalog No. 502973072
Encompass_Preferred
A plant-derived steroidal triterpene that induces cell cycle arrest or apoptosis in a range of cancer cell lines; inhibits the growth of several breast cancer cell lines (IC50s = 18-50 nM); suppresses the growth of tumors developed from MDA-MB-231 and 4T-1 breast cancer cells in mice
Catalog No. 50-297-3072 Supplier Cayman Chemical Supplier No. 1482010 MG
A plant-derived steroidal triterpene that induces cell cycle arrest or apoptosis in a range of cancer cell lines; inhibits the growth of several breast cancer cell lines (IC50s = 18-50 nM); suppresses the growth of tumors developed from MDA-MB-231 and 4T-1 breast cancer cells in mice