Learn More
Medchemexpress LLC Cucurbitacin B | 6199-67-3 | MFCD07778083 | 98.2% | 558.70 g/mol | C32H46O8 | 10 MG

Supplier: Medchemexpress LLC HYN0416R10MG
Cucurbitacin B analytical standard is a reference material of the tetracyclic triterpenoid Cucurbitacin B intended for research and analytical applications, including qualitative and quantitative method development by HPLC, GC, and MS. Supplied as a solid (white to off-white), it is provided in a small quantity for assay verification and calibration.
- Analytical standard for HPLC, GC, and MS
- High purity (~98.2%) suitable for quantitative assays
- Molecular formula C32H46O8 and molecular weight 558.70 g/mol
- White to off-white solid derived from plant sources
- Packaged as a 10 mg quantity for analytical use
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.