Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cucurbitacin B (Standard) | 6199-67-3 | 558.70 | 100 MG
SDP

Catalog No. 5000390284
Encompass_Preferred

Cucurbitacin B (Standard) is the analytical standard of Cucurbitacin B, intended for research and analytical applications. It is a highly oxidized tetracyclic triterpenoid and is orally active. It inhibits tumor cell growth, migration, invasion, and induces cell cycle arrest and apoptosis. It also possesses potent anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities.

  • Application: The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants (Cucurbitaceae, Trichosanthes cucumerina L.)
  • Endogenous metabolite

Catalog No. 50-003-90284 Supplier Medchemexpress LLC Supplier No. HYN0416R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Cucurbitacin B (Standard) is the analytical standard of Cucurbitacin B, intended for research and analytical applications. It is a highly oxidized tetracyclic triterpenoid and is orally active. It inhibits tumor cell growth, migration, invasion, and induces cell cycle arrest and apoptosis. It also possesses potent anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities.

  • Application: The compound is an analytical standard used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Terpenoids, Triterpenes
  • Initial source: Plants (Cucurbitaceae, Trichosanthes cucumerina L.)
  • Endogenous metabolite

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.