Learn More
Medchemexpress LLC Cucurbitacin B (Standard) | 6199-67-3 | 558.70 | 25 MG
SDP
Catalog No. 5000389333
Encompass_Preferred
Cucurbitacin B (Standard) is an analytical standard of Cucurbitacin B for research and analytical applications. This bioactive tetracyclic triterpenoid inhibits tumor cell growth, migration, invasion, and induces cell apoptosis. It is commonly used in qualitative, quantitative, and methodological research experiments as a reference standard.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell apoptosis
  • Possesses anti-inflammatory activity
  • Acts as an antioxidant
  • Has antiviral properties
  • Exhibits hypoglycemic effects
  • Provides hepatoprotective benefits
  • Offers neuroprotective activity
  • Used as a reference standard in HPLC, GC, and MS
Catalog No. 50-003-89333 Supplier Medchemexpress LLC Supplier No. 25MGHYN0416R
Cucurbitacin B (Standard) is an analytical standard of Cucurbitacin B for research and analytical applications. This bioactive tetracyclic triterpenoid inhibits tumor cell growth, migration, invasion, and induces cell apoptosis. It is commonly used in qualitative, quantitative, and methodological research experiments as a reference standard.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell apoptosis
  • Possesses anti-inflammatory activity
  • Acts as an antioxidant
  • Has antiviral properties
  • Exhibits hypoglycemic effects
  • Provides hepatoprotective benefits
  • Offers neuroprotective activity
  • Used as a reference standard in HPLC, GC, and MS