Learn More
Medchemexpress LLC Cucurbitacin B (standard) | 6199-67-3 | 558.70 | 5 MG
SDP
Catalog No. 5000389919
Encompass_Preferred
Cucurbitacin B (Standard) is the analytical standard of Cucurbitacin B, intended for research and analytical applications. It is a highly oxidized tetracyclic triterpenoid and is orally active. Cucurbitacin B has been shown to inhibit tumor cell growth, migration, and invasion, induce cell cycle arrest and apoptosis, and possess potent anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities. It is used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell cycle arrest and apoptosis
  • Possesses anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-89919 Supplier Medchemexpress LLC Supplier No. HYN0416R5MG
Cucurbitacin B (Standard) is the analytical standard of Cucurbitacin B, intended for research and analytical applications. It is a highly oxidized tetracyclic triterpenoid and is orally active. Cucurbitacin B has been shown to inhibit tumor cell growth, migration, and invasion, induce cell cycle arrest and apoptosis, and possess potent anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities. It is used as a reference standard in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell cycle arrest and apoptosis
  • Possesses anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)