Learn More
Medchemexpress LLC Cucurbitacin B (Standard) | 6199-67-3 | 558.70 | 50 MG
SDP
Catalog No. 5000389983
Encompass_Preferred
Cucurbitacin B (Standard) is an analytical reference standard intended for research and analytical applications. This orally active, highly oxidized tetracyclic triterpenoid inhibits tumor cell growth, migration, and invasion, and induces cell apoptosis. It also demonstrates anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities. It is widely used in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell apoptosis
  • Exhibits anti-inflammatory activity
  • Offers antioxidant properties
  • Demonstrates antiviral effects
  • Has hypoglycemic activity
  • Provides hepatoprotective benefits
  • Shows neuroprotective activities
  • Used as a reference standard for assays
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-003-89983 Supplier Medchemexpress LLC Supplier No. HYN0416R50MG
Cucurbitacin B (Standard) is an analytical reference standard intended for research and analytical applications. This orally active, highly oxidized tetracyclic triterpenoid inhibits tumor cell growth, migration, and invasion, and induces cell apoptosis. It also demonstrates anti-inflammatory, antioxidant, antiviral, hypoglycemic, hepatoprotective, and neuroprotective activities. It is widely used in qualitative, quantitative, and methodological research experiments including HPLC, GC, and MS.
  • Inhibits tumor cell growth, migration, and invasion
  • Induces cell apoptosis
  • Exhibits anti-inflammatory activity
  • Offers antioxidant properties
  • Demonstrates antiviral effects
  • Has hypoglycemic activity
  • Provides hepatoprotective benefits
  • Shows neuroprotective activities
  • Used as a reference standard for assays
  • Suitable for HPLC, GC, and MS research experiments