Learn More
Selleck Chemical LLC Cucurbitacin D-E0701-1MG
SDP
Catalog No. 503136888
Encompass_Preferred
Cucurbitacin D is an active component in Cucurbita texana disrupts maturation of HSP90 and interactions between Hsp90 and two co-chaperones Cdc37 and p23
Catalog No. 50-313-6888 Supplier Selleck Chemical LLC Supplier No. E07011MG
Cucurbitacin D is an active component in Cucurbita texana disrupts maturation of HSP90 and interactions between Hsp90 and two co-chaperones Cdc37 and p23