Learn More
Medchemexpress LLC Cucurbitacin E | 18444-66-1 | 556.69 | 1 MG
SDP
Catalog No. 5000389920
Encompass_Preferred
Cucurbitacin E is a natural compound sourced from Cucurbitaceae plants. It significantly suppresses the activity of the cyclin B1/CDC2 complex and is intended for research use only. This compound exhibits growth inhibition in various cancer cell lines and reduces obesity in mice by targeting the JAK-STAT5 signaling pathway.
  • Natural compound from Cucurbitaceae plants.
  • Significantly suppresses the activity of the cyclin B1/CDC2 complex.
  • Exhibits growth inhibition in various cancer cell lines including human prostate, CNS, breast, colon, HL-60, HSC-T6, HT-1080, HeLa, HepG2, JY, MCF7, and U-937 cells.
  • Induces morphological changes in primary colon cancer cells and inhibits tumor growth by arresting the cell cycle in the G2/M phase.
  • Reduces obesity and related metabolic dysfunction in mice.
Catalog No. 50-003-89920 Supplier Medchemexpress LLC Supplier No. HYN04171MG
Cucurbitacin E is a natural compound sourced from Cucurbitaceae plants. It significantly suppresses the activity of the cyclin B1/CDC2 complex and is intended for research use only. This compound exhibits growth inhibition in various cancer cell lines and reduces obesity in mice by targeting the JAK-STAT5 signaling pathway.
  • Natural compound from Cucurbitaceae plants.
  • Significantly suppresses the activity of the cyclin B1/CDC2 complex.
  • Exhibits growth inhibition in various cancer cell lines including human prostate, CNS, breast, colon, HL-60, HSC-T6, HT-1080, HeLa, HepG2, JY, MCF7, and U-937 cells.
  • Induces morphological changes in primary colon cancer cells and inhibits tumor growth by arresting the cell cycle in the G2/M phase.
  • Reduces obesity and related metabolic dysfunction in mice.