Learn More
Medchemexpress LLC Cucurbitacin E | 18444-66-1 | 556.69 | 1 ML
SDP
Catalog No. 5000389132
Encompass_Preferred
Cucurbitacin E is a natural compound derived from Cucurbitaceae plants. It significantly suppresses the activity of the cyclin B1/CDC2 complex and exhibits antitumor activity.
  • Natural compound
  • Suppresses cyclin B1/CDC2 complex activity
  • Used in cancer research
  • Explored for antitumor activity against colorectal cancer cells
  • Induces morphological changes in primary colon cancer cells
  • Inhibits tumor growth by arresting the cell cycle
Catalog No. 50-003-89132 Supplier Medchemexpress LLC Supplier No. HYN041710MM/1ML
Cucurbitacin E is a natural compound derived from Cucurbitaceae plants. It significantly suppresses the activity of the cyclin B1/CDC2 complex and exhibits antitumor activity.
  • Natural compound
  • Suppresses cyclin B1/CDC2 complex activity
  • Used in cancer research
  • Explored for antitumor activity against colorectal cancer cells
  • Induces morphological changes in primary colon cancer cells
  • Inhibits tumor growth by arresting the cell cycle